Learn More
Cytiva Amersham™ Hyperfilm™ ECL
Use with all blue and green light chemiluminescent systems. Cytiva Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
Brand:  Cytiva 28-9068-38
308.68 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 10479214
  • £365.00 / 50 sheets
Explore more special offers
Description
Description
  •  Antistatic coating reduces risk of visible artifacts
  • Clear base offers high contrast for easy reading of images
  • Suitable for use with all blue and green light chemiluminescent systems
  • Film can be processed in automatic processors or manually using most common X-ray film developers and fixers
  • Chemiluminescent detection in Western, Southern, and Northern blots
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amersham Hyperfilm ECL
Chemiluminescent detection in Western, Southern, and Northern blots.
4°C
20.3 x 25.4 cm (8 x 10 in.)
50 Sheets/Box