Learn More
Cytiva Amersham™ Hyperfilm™ MP
Use in automatic processors or manually in tanks and trays using most common X-ray film developers and fixers. Cytiva Life Sciences™ Amersham™ Hyperfilm™ MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti scratch layer.
Brand:  Cytiva 28-9068-46
468.22 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
 View more versions of this product
Product Code. 10627265
  • £539.00 / 100 sheets
Explore more special offers
Description
Description
  • High sensitivity for low concentration targets
  • Optimized for direct and indirect autoradiography with 3H, 14C, 32P, 33P, 35S, 125I
  • Clear background for excellent contrast and improved band visibility.
  • Publication-quality images
  • High resolution at an economical price
  • Easy to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8 x 10 in.
32P, 33P, 125I Southern, Northern, and Western blots. 32S, 33P, 35P sequencing. Fluorography of 3H, 14C, and 35S
100 Sheets/Box, Single film bag