Learn More
Cytiva Amersham™ Megaprime™ DNA Labeling System
For labeling DNA probes to specific activities >1 x 109dpm/μg in 10 minutes
Marke:  Cytiva RPN1604
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 10025174
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen »
Entdecken Sie weitere Sonderangebote
Beinhaltet: Primer solution; Labeling buffer; Reaction buffer; Enzyme; Standard DNA solution; Carrier DNA solution
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
Megaprime™ DNA Labeling Systems are used for rapid, random-prime labeling from as little as 25ng substrate DNA.
  • Features cloned Klenow fragment and random nonamers in the random-prime labeling technique
  • No need to remove unincorporated nucleotides before hybridization
  • Protocols for labeling directly from low melting point agarose
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
-15°C to -30°C
Primer solution 150μL; Labeling Buffer dATP, dGTP and dTTP in Tris HCl pH7.5, 2-mercaptoethanol and MgCl2 Nucleotide solutions: dATP, cCTP, dGTP, dTTP in Tris HCl pH 8, 0.5mM EDTA 120μL, Reaction Buffer A 10 x concentra
Megaprime DNA Labeling System, dNTP (for use with any radioactively labeled nucleotide)
Membrane hybridization
30
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen
Sonderangebote
Sonderangebote