Learn More
Cytiva Amersham™ Reinforced Nitrocellulose Blotting Membrane
Nitrocellulose membrane, with a 0.45 µm pore size, is suitable for proteins of a wide range of molecular weights.
Brand:  Cytiva 10600124
80.64 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Product Code. 15255928
  • £87.53 / 10 sheets
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Low background and excellent signal-to-noise ratios.
  • Compatible with a variety of labeling and detection systems.
  • High binding capacity.
  • Suitable for proteins of a wide range of molecular weights.
  • Proprietary nitrocellulose formula gives a long shelf life of bound proteins.
  • Available in popular precut sizes for your application.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blotting Membrane
Protein
1 Each
For western, southern and northern blotting applications
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • ||Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose