missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ Streptavidin-Fluor Conjugates

Use for in situ hybridization and flow cytometry. Cytiva Life Sciences™ Amersham™ Streptavidin-Fluor Conjugates consist of streptavidin conjugated to a range of fluorescent dyes.

Brand:  Cytiva PA45001

209.85 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.


 View more versions of this product

Product Code. 10004594

  • £254.00 / 1mg
Estimated Shipment: 14-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • High specificity reagents

Applications:

in situ hybridization, flow cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

This product is supplied as a lyophilized solid in Phosphate-buffered Saline (0.02M Potassium Phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) Bovine Serum Albumin and 0.1% (w/v) Sodium Azide, reconstitute with 1mL of deionized Water, dispense into suitable aliquots and freeze, avoid repeated freeze-thaw cycles
1 mg
Lyophilized solid in phosphate-buffered saline (0.02M potassium phosphate, 0.15M NaCl) pH 7.2, containing 1%(w/v) bovine serum albumin and 0.1% (w/v) sodium azide, reconstitute with 1mL of deionized water, dispense into suitable aliquots and freeze
Suitable for use in the Fluorescent analysis of Biotinylated samples
Cy5-Streptavidin
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.