missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ TE 70 and TE 70 PWR Semi-Dry Transfer Unit
Electrotransfer proteins from polyacrylamide gels up to 14 x 16cm (TE 77 PWR) and 21 x 26cm (TE 77) in size in less than one hour
£1125.00 - £1426.00
Product Code | Brand | Includes | Model | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Model | Price | Quantity & Availability | ||||
10468264
|
Cytiva
11-0013-41 |
25 sheets of blotting paper, 50 sheets of cellophane, 2 solid masks, masks for 7 x 8 cm and 14 x 16 cm gels. | TE 70 PWR, with Integral power supply | N/A | |||||
10420550
|
Cytiva
TE70 |
25 sheets blotting paper, 50 sheets cellophane, 2 solid masks, masks for 7 x 8 cm and 14 x 16 cm gels. | TE 70 | N/A | |||||
Description
- Long-lasting, perforated platinum-titanium and stainless-steel electrodes give uniform, contamination-free transfers
- Requires minimal buffer, only enough to saturate blotting papers and membranes
- Uses low current (0.5A) and voltage (30V) for transfers: safer and more economical
- Suitable for gels up to 14 × 16cm and 21 × 26cm
- Tested for use with Amersham™ ECL Western Blotting reagents
- Unique automatic stopping feature; transfer is stopped when buffer gets depleted, preventing overheating
Specifications
14 x 16 cm | |
38 cm | |
9 cm |