missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-AMG 487, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
(±)-AMG 487 is an antagonist of CXCR3; inhibits binding of 125I-IP-10 and 125I-ITAC to CXCR3 (IC50 values are 8.0 and 8.2 nM respectively). Inhibits CXCR3-mediated cell migration by the chemokines IP-10, ITAC and MiG in vitro (IC50 values are 8, 15 and 36 nM respectively). Also shown to inhibit lung metastasis in a mouse model of metastatic breast cancer.
This product is racemic.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 603.59 |
Percent Purity | >98% |
Chemical Name or Material | (±)-AMG 487 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction