missing translation for 'onlineSavingsMsg'
Learn More

(±)-AMG 487, Tocris Bioscience™

Product Code. 30001673
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

CXCR3 antagonist; inhibits cell migration and metastasis

(±)-AMG 487 is an antagonist of CXCR3; inhibits binding of 125I-IP-10 and 125I-ITAC to CXCR3 (IC50 values are 8.0 and 8.2 nM respectively). Inhibits CXCR3-mediated cell migration by the chemokines IP-10, ITAC and MiG in vitro (IC50 values are 8, 15 and 36 nM respectively). Also shown to inhibit lung metastasis in a mouse model of metastatic breast cancer.

This product is racemic.

Chemical Identifiers

CAS 947536-03-0
Molecular Formula C32H28F3N5O4
Molecular Weight (g/mol) 603.602
InChI Key WQTKNBPCJKRYPA-UHFFFAOYSA-N
Synonym n-1r-1-3-4-ethoxyphenyl-4-oxopyrido 2,3-d pyrimidin-2-yl ethyl-n-pyridin-3-ylmethyl-2-4-trifluoromethoxy phenyl acetamide, n-1-3-4-ethoxyphenyl-3,4-dihydro-4-oxopyrido 2,3-d pyrimidin-2-yl ethyl-n-3-pyridinylmethyl-4-trifluoromethoxy benzeneacetamide
PubChem CID 10282265
IUPAC Name N-[1-[3-(4-ethoxyphenyl)-4-oxopyrido[2,3-d]pyrimidin-2-yl]ethyl]-N-(pyridin-3-ylmethyl)-2-[4-(trifluoromethoxy)phenyl]acetamide
SMILES CCOC1=CC=C(C=C1)N2C(=O)C3=C(N=CC=C3)N=C2C(C)N(CC4=CN=CC=C4)C(=O)CC5=CC=C(C=C5)OC(F)(F)F

Specifications

Quantity 50 mg
Formula Weight 603.59
Percent Purity >98%
Chemical Name or Material (±)-AMG 487
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.