missing translation for 'onlineSavingsMsg'
Learn More
AMG 9810, Tocris Bioscience™ Product Code.: 30001298

AMG 9810, Tocris Bioscience™

Product Code. 30001298
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001298

Brand: Tocris Bioscience 2316/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective, competitive antagonist of TRPV1

Potent and selective, competitive vanilloid TRPV1 receptor antagonist (IC50 = 17 nM). Inhibits capsaicin-, proton-, heat- and endogenous ligand-induced activation of human and rat recombinant TRPV1 receptors. Displays antihyperalgesic properties in a rat model of inflammatory pain.

Chemical Identifiers

CAS 545395-94-6
Molecular Formula C21H23NO3
Molecular Weight (g/mol) 337.419
InChI Key GZTFUVZVLYUPRG-IZZDOVSWSA-N
Synonym unii-182hij2d7f, 2e-3-4-tert-butylphenyl-n-2,3-dihydro-1,4-benzodioxin-6-yl prop-2-enamide, 2e-n-2,3-dihydro-1,4-benzodioxin-6-yl-3-4-1,1-dimethylethyl phenyl-2-propenamide, trans-cinnamide, d0xo2g, e-3-4-t-butylphenyl-n-2,3-dihydrobenzo b 1,4 dioxin-6-yl acrylamide, 3-4-t-butylphenyl-n-2,3-dihydrobenzo b 1,4 dioxin-6-yl acrylamide, amg hplc , powder, e-3-4-tert-butylphenyl-n-2,3-dihydro-1,4-benzodioxin-6-yl prop-2-enamide
PubChem CID 680502
IUPAC Name (E)-3-(4-tert-butylphenyl)-N-(2,3-dihydro-1,4-benzodioxin-6-yl)prop-2-enamide
SMILES CC(C)(C)C1=CC=C(C=C1)C=CC(=O)NC2=CC3=C(C=C2)OCCO3

Specifications

Quantity 10 mg
Formula Weight 337.42
Percent Purity >98%
Chemical Name or Material AMG 9810
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
AMG 9810, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.