Learn More
EVOS™ EVOS™ Onstage Vessel Holder, Two 25 mL Flask
Brand:  EVOS™ AMEPVH034
Code : NEW
Additional Details : Weight : 1.00000kg
Product Code. 15215013
  • £302.00 / Each
Estimated Shipment: 12-08-2024
to see stock.
Description
Description
For use with EVOS FL Auto Digital Inverted Fluorescence System (Cat. No. 12-463-377).
Onstage Vessel Holders
  • Accommodate a variety of flasks, plates and dishes
  • Maximize your choice of cell culture vessels when conducting time-lapse imaging experiments using the EVOS FL Auto Imaging System equipped with onstage incubator
  • Allow a perfect fit of your microwell plates or cell culture flasks/dishes to the master plate of the onstage incubator for increased precision in sample alignment
  • With a snug and secure fit, yet readily switched, these vessel holders make time-lapse imaging convenient and easy
Specifications
Specifications
Vessel Holder
EVOS™ FL Auto Imaging System
1/Ea.
EVOS
Holds two 25 mL T-flasks