Learn More
EVOS™ EVOS™ Onstage Vessel Holder, Multi-Well Plates
Brand: EVOS™ AMEPVH028
Code : NEW
Additional Details : Weight : 1.00000kg
Description
For use with EVOS FL Auto Digital Inverted Fluorescence System (Cat. No. 12-463-377).
Onstage Vessel Holders
- Accommodate a variety of flasks, plates and dishes
- Maximize your choice of cell culture vessels when conducting time-lapse imaging experiments using the EVOS FL Auto Imaging System equipped with onstage incubator
- Allow a perfect fit of your microwell plates or cell culture flasks/dishes to the master plate of the onstage incubator for increased precision in sample alignment
- With a snug and secure fit, yet readily switched, these vessel holders make time-lapse imaging convenient and easy
Specifications
Vessel Holder | |
EVOS™ FL Auto Imaging System | |
1 each |
EVOS | |
Holds one multi-well plate |
For Research Use Only. Not for use in diagnostic procedures.