missing translation for 'onlineSavingsMsg'
Learn More

EVOS™ EVOS™ Vessel Holder, SPL T-75 flask

Product Code. 11977325
Click to view available options
Quantity:
1 Holder
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11977325

Brand: EVOS™ AMEPVH012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Securely holds one SPL T-75 flask

  • For use with EVOS and Micromaster inverted microscopes
  • Holds one SPL T-75 flask
TRUSTED_SUSTAINABILITY

Specifications

Type Vessel Holder
Product Line EVOS
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Format one, SPL T-75 flask
Holds 1 SPL T-75 Flask
Quantity 1 Holder

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.