missing translation for 'onlineSavingsMsg'
Learn More

Merck Amicon™ Stirred Cells

Amicon stirred cells concentrate samples rapidly but gently, using magnetic stirring to minimize concentration polarization and shear stress-induced denaturation.

Brand:  Merck UFSC05001

 View more versions of this product

Product Code. 15572061

  • £1990.00 / Each
Estimated Shipment: 08-05-2025
to see stock.
Explore more special offers
Add to basket

Alert:

Pressure Limitation. Do not operate the stirred cell above 5.2 bar (75 psi).

This item is not returnable. View return policy

Description

Description

This Amicon stirred cell model provides high flow rates with solutions up to 10% macrosolute concentration. Capable of rapid concentration, or salt removal followed by concentration in the same unit.

Services

One year

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Ultracel™, Biomax™, and Durapore™ membrane discs for use in stirred cells
Amicon™ Stirred Cell 50 mL
50 mL
Amicon Stirred Cell 50 mL
Concentration, Buffer Exchange, Desalting
Ultrafiltration Equipment
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Merck Amicon™ Stirred Cells > 50mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.