missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Ultra-15 Centrifugal Filter Units
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
£209.00 - £2010.00
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | High-recovery Ultracel-PL Regenerated Cellulose |
Sterility | Non-sterile |
Filtration Area | 7.6 cm2 |
For Use With (Application) | Protein purification, protein concentration and desalting |
Product Code | Brand | Diameter (Metric) | MWCO | Volume (Metric) | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | MWCO | Volume (Metric) | Quantity | Price | Quantity & Availability | ||
10705884
|
Merck
UFC900308 |
29.7 mm | 3 kDa | 15 mL | 8Pack |
£210.00
Pack of 8 |
N/A | ||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10092423
|
Merck
UFC900396 |
29.7 mm | 3 kDa | 15 mL | 96 Pack |
£1490.00
Pack of 96 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|
10608072
|
Merck
UFC901008 |
29.7 mm | 10 kDa | 15 mL | 8Pack |
£222.00
Pack of 8 |
In Stock
Log in to see stock. |
||
10781543
|
Merck
UFC901024 |
29.7 mm | 10 kDa | 15 mL | 24Pack |
£524.00
Pack of 24 |
In Stock
Log in to see stock. |
||
10026941
|
Merck
UFC901096 |
29.7 mm | 10 kDa | 15 mL | 96Pack |
£1915.00
Pack of 96 |
In Stock
Log in to see stock. |
||
10581342
|
Merck
UFC903008 |
29.7 mm | 30 kDa | 15 mL | 8Pack |
£237.00
Pack of 8 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||
10311882
|
Merck
UFC903024 |
29.7 mm | 30 kDa | 15 mL | 24Pack |
£496.00
Pack of 24 |
In Stock
Log in to see stock. |
||
10036941
|
Merck
UFC903096 |
29.7 mm | 30 kDa | 15 mL | 96Pack |
£1970.00
Pack of 96 |
In Stock
Log in to see stock. |
||
10550443
|
Merck
UFC905008 |
29.7 mm | 50 kDa | 15 mL | 8Pack |
£235.00
Pack of 8 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||
10413892
|
Merck
UFC905096 |
29.7 mm | 50 kDa | 15 mL | 96Pack |
£1525.00
Pack of 96 |
Estimated Shipment: 01-11-2024
Log in to see stock. |
||
10188431
|
Merck
UFC910024 |
29.7 mm | 100 kDa | 15 mL | 24Pack |
£540.77
Pack of 24 |
In Stock
Log in to see stock. |
||
10447871
|
Merck
UFC910096 |
29.7 mm | 100 kDa | 15 mL | 96Pack |
£2010.00
Pack of 96 |
N/A | ||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
10403892
|
Merck
UFC900324 |
29.7 mm | 3 kDa | 15 mL | 24Pack |
£524.00
Pack of 24 |
In Stock
Log in to see stock. |
|
10250482
|
Merck
UFC910008 |
29.7 mm | 100 kDa | 15 mL | 8Pack |
£209.00
Pack of 8 |
In Stock
Log in to see stock. |
||
10661043
|
Merck
UFC905024 |
29.7 mm | 50 kDa | 15 mL | 24Pack |
£540.77
Pack of 24 |
In Stock
Log in to see stock. |
||
Description
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
- High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
- Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
- Heat-sealed membrane eliminates possibility of downstream extractables
- Universal rotor compatibility
- High retentate recovery of >90%
- High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
- Easy sample monitoring due to translucent housing and volume gradations
- Direct pipetter sample access eliminates processing step to recover concentrate
- Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
- Purification of macromolecular components found in tissue culture extracts or cell lysates
- Concentration of dilute or prepurified proteins from column eluents
- Desalting and buffer exchange
Specifications
Single-use | |
Non-sterile | |
Protein purification, protein concentration and desalting | |
Two microcentrifuge tubes | |
15 mL | |
29.7 mm |
High-recovery Ultracel-PL Regenerated Cellulose | |
7.6 cm2 | |
Regenerated Cellulose (RC) | |
Centrifugal Filters | |
121 mm |