missing translation for 'onlineSavingsMsg'
Learn More

Merck Amicon™ Ultra-4 Centrifugal Filter Units

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

Brand:  Merck UFC803096

642.66 GBP valid until 2024-12-31
Use promo code "23269" to get your promotional price.



Code : 72

Additional Details : Weight : 1.45800kg

 View more versions of this product

Product Code. 10210342

  • £1180.00 / Pack of 96
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Includes: Two microcentrifuge tubes

Description

Description

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange
Specifications

Specifications

Single-use
30 kDa
3 cm2
Regenerated Cellulose (RC)
96Pack
4 mL
22.7 mm
High-recovery Ultracel-PL Regenerated Cellulose
Non-sterile
Protein purification, protein concentration and desalting
Two microcentrifuge tubes
Centrifugal Filters
124 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers