missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Amido Black 10B, 0.2% v/v soln. in 5% acetic acid

Catalog Number 15420227
Click to view available options
Quantity:
500 mL
1 L
This item is not returnable. View return policy
1064-48-8
C22H14N6Na2O9S2
616.487
MFCD00004017
HKBVRFLHNUEVRO-UHFFFAOYSA-L
Acid black 1; C.I. 20470
44134531
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
This item is not returnable. View return policy
1064-48-8
C22H14N6Na2O9S2
616.487
MFCD00004017
HKBVRFLHNUEVRO-UHFFFAOYSA-L
Acid black 1; C.I. 20470
44134531
disodium;4-amino-3-[(4-nitrophenyl)diazenyl]-5-oxo-6-(phenylhydrazinylidene)naphthalene-2,7-disulfonate
C1=CC=C(C=C1)NN=C2C(=CC3=CC(=C(C(=C3C2=O)N)N=NC4=CC=C(C=C4)[N+](=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration 0.2% v/v soln. in 5% acetic acid
Beilstein 5374263
Formula Weight 616.49
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Amido Black 10B

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'