missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Amido Black 10B, Electrophoresis Reagent
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Description
Used to stain proteins in polyacrylamide gels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Beilstein | 5374263 |
Formula Weight | 616.49 |
Packaging | Plastic bottle |
Sensitivity | Hygroscopic |
Quantity | 25 g |
Physical Form | Powder |
Chemical Name or Material | Amido Black 10B |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'