Learn More
Thermo Scientific™ Amies Agar Gel Mini Tip: Firm Applicator Wire
Simplify the transportation of eye, nasopharyngeal, male urethral swab and pediatric swabs with these Amies Agar Gel Transport Swabs.
Brand: Thermo Scientific™ TS0012A
Code : 50
Additional Details : Weight : 0.50000kg
Description
Simplify the transportation of eye, nasopharyngeal, male urethral swab and pediatric sampling with Thermo Scientific™ Oxoid™ Amies Agar Gel Mini Tip: Firm Applicator Wire. The narrow dimension of the swab shaft and tip provides a more practical device for some sampling.
Ensure optimal sample collection and recovery using Amies Agar Gel Mini Tip: Firm Applicator Wire.
- Contains media for the maintenance of bacterial sample during transport to the laboratory.
- Each batch of product is tested for performance using a wide range of aerobic and anaerobic pathogens to ensure adequate recoveries.
- Lot number/expiry date and tamper-evident tube ensure complete traceability and sterility.
- “Venturi” Hour-Glass Tube Design prevents disintegration of gel column and eliminates undesirable air pockets, bubbles and breaks in gel which may be harmful to fastidious bacteria
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Transport Swab | |
50/Pk., 500/Cs. | |
5°C to 25C°; do not freeze | |
Minitip |
Amies Agar Gel Mini Tip: Firm Applicator Wire | |
Aluminum Wire | |
Rayon |