Learn More
Amikacin disulfate, MedChemExpress
MedChemExpress Amikacin disulfate (BAY 41-6551 dissulfate) is an aminoglycoside antibiotic and a semisynthetic analog of kanamycin.

£93.17 - £375.10
Specifications
Chemical Name or Material | Amikacin disulfate |
---|---|
CAS | 39831-55-5 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C22H47N5O21S2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16440968
![]() |
Medchem Express
HY-B0509B |
5 g |
£375.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16412209
![]() |
Medchem Express
HY-B0509B |
1 g |
£93.17
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Amikacin disulfate is bactericidal, acting directly on the 30S and 50S bacerial ribosomal subunits to inhibit protein synthesis. Amikacin disulfate is very active against most Gram-negative bacteria including gentamicin- and tobramycin-resistant strains. Amikacin disulfate also inhibits the infections caused by susceptible Nocardia and nontuberculous mycobacteria.Specifications
Amikacin disulfate | |
White | |
C22H47N5O21S2 | |
H2O : 100 mg/mL (127.92 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) | |
781.76 | |
98.0% | |
Solid |
39831-55-5 | |
Research | |
BAY 41-6551 disulfate | |
OS(O)(=O)=O.OS(O)(=O)=O.O[C@@H]1[C@H]([C@@H](C[C@H](N)[C@H]1O[C@@]([C@@H]([C@@H](O)[C@@H]2O)O)([H])O[C@@H]2CN)NC([C@@H](O)CCN)=O)O[C@@]([C@@H]([C@@H](N)[C@@H]3O)O)([H])O[C@@H]3CO | |
781.76 | |
Research | |
COVID-19-immunoregulation |
Safety and Handling
H315∣H319∣H335∣H350∣H360
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.