missing translation for 'onlineSavingsMsg'
Learn More

Amikacin (From Kanamycin A, White Crystalline Powder), Fisher BioReagents

Product Code. 10134924
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass
This item is not returnable. View return policy
This item is not returnable. View return policy
  • An antibiotic that serves as a selective agent to isolate transformed bacteria
  • Amikacin functions as an indirect inhibitor of protein synthesis in prokaryotes by binding to the 30S subunit of ribosomes
  • Useful as a selective agent to isolate bacteria that have been transformed with plasmid containing the antibiotic-resistant gene
  • Intended for research purposes only; not for manufacturing or commercial use
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37517-28-5
Molecular Formula C22H43N5O13
Molecular Weight (g/mol) 585.608
InChI Key LKCWBDHBTVXHDL-RMDFUYIESA-N
Synonym Lukadin
PubChem CID 37768
ChEBI CHEBI:2637
IUPAC Name (2S)-4-amino-N-[(1R,2S,3S,4R,5S)-5-amino-2-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide
SMILES C1C(C(C(C(C1NC(=O)C(CCN)O)OC2C(C(C(C(O2)CO)O)N)O)O)OC3C(C(C(C(O3)CN)O)O)O)N

Specifications

Type Amikacin
Physical Form Solid
pH 9.5 to 11.5
Melting Point 214°C
Merck Index 15, 400
Packaging Amber Glass
Potency > 900μg/mg
Solubility Information completely soluble in water
Formula Weight 585.61
Color White
Quantity 250 mg
Chemical Name or Material Amikacin, From Kanamycin A
Show More Show Less
Product Identifier
  • Amikacin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.