missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Amlexanox
GREENER_CHOICE

Product Code. 30001599
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001599

Brand: Tocris Bioscience™ 4857/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of TBK

Selective inhibitor of TANK-binding kinase 1 (TBK1) and IKKε (IC50 values are ∽1-2 μM). Displays no effect on IKKα or IKKβ at these concentrations. Reversibly lowers weight, increases insulin sensitivity, and reduces inflammation and steatosis in three mouse models of obesity. Exhibits antiallergic activity; inhibits the release of histamine from rat mast cells. Also binds to Hsp90 and inhibits C-terminal chaperone activity in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68302-57-8
Molecular Formula C16H14N2O4

Specifications

Quantity 10 mg
Formula Weight 298.29
Purity >99%
Molecular Weight (g/mol) 298.30
Chemical Name or Material Amlexanox
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.