missing translation for 'onlineSavingsMsg'
Learn More
Amlexanox, Tocris Bioscience™ Product Code.: 30001620

Amlexanox, Tocris Bioscience™

Product Code. 30001620
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001620

Brand: Tocris Bioscience 4857/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of TBK

Selective inhibitor of TANK-binding kinase 1 (TBK1) and IKKε (IC50 values are ∽1-2 μM). Displays no effect on IKKα or IKKβ at these concentrations. Reversibly lowers weight, increases insulin sensitivity, and reduces inflammation and steatosis in three mouse models of obesity. Exhibits antiallergic activity; inhibits the release of histamine from rat mast cells. Also binds to Hsp90 and inhibits C-terminal chaperone activity in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68302-57-8
Molecular Formula C16H14N2O4
Molecular Weight (g/mol) 298.30
MDL Number MFCD00864790
InChI Key SGRYPYWGNKJSDL-UHFFFAOYSA-N
Synonym amlexanox, aphthasol, amoxanox, 2-amino-7-isopropyl-5-oxo-5h-chromeno 2,3-b pyridine-3-carboxylic acid, amlenanox, solfa, amlexanoxo, amlexanoxum, amlexanoxum latin, amlexanoxo spanish
PubChem CID 2161
ChEBI CHEBI:31205
IUPAC Name 2-amino-5-oxo-7-(propan-2-yl)-5H-chromeno[2,3-b]pyridine-3-carboxylic acid
SMILES CC(C)C1=CC=C2OC3=NC(N)=C(C=C3C(=O)C2=C1)C(O)=O

Specifications

Quantity 50 mg
Formula Weight 298.29
Percent Purity >99%
Chemical Name or Material Amlexanox
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amlexanox, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.