missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 368380010

343.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



 View more versions of this product

Product Code. 10127253

  • £344.72 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467900 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number368380010,368381000
Lot NumberA0460989 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog Number368380010,368381000
Lot NumberA0450126 Certificate TypeCertificate of Analysis Date26/02/2024 Catalog Number368380010,368381000
Lot NumberA0457878 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number368380010,368381000
Lot NumberA0453619 Certificate TypeCertificate of Analysis Date14/12/2023 Catalog Number368380010,368381000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonia, 0.5M solution in 1,4-dioxane, AcroSeal™, Thermo Scientific Chemicals > 1L; AcroSeal glass bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only