Learn More
Ammonia 10 M NaOH Ionic Strength Adjuster Solution, Reagecon
Keeps ionic strength of standard and test high to eliminate matrix effects and suppress or eliminate interfering ions.
Brand: Reagecon ISANH5
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 1824 Chem class : 8 Pack group : II
Description
- High ionic strength
- Aqueous matrix
- Used as an additive when using an ion selective electrode (ISE) for the measurement of ammonia.
- Suppresses interfering ions, eliminates matrix, hysteresis or liquid junction potentials.
- Eliminates or reduces error due to temperature co-efficient of variation between sample and calibration standard.
- Certified and has guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Physical form: Liquid | |
HDPE Bottle GL45 | |
Ion Selective Electrode |
Ammonia 10M NaOH | |
500 mL |
Safety and Handling
- AMMONIA ISE IONIC STRENGTH
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium hydroxide