Learn More
Ammonia, 2M solution in methanol, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 368391000
54.64 GBP valid until 2024-12-31
Use promo code "21665" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ammonia | |
7664-41-7, 67-56-1 | |
NH3 | |
MFCD00011418 | |
0.7870g/mL | |
ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol | |
QGZKDVFQNNGYKY-UHFFFAOYSA-N | |
N | |
17.03 | |
222 | |
17.03 | |
0.787 |
Colorless | |
H3N | |
1.90 to 2.20M | |
05,15 | |
15, 488 | |
Solubility in water: soluble. Other solubilities: soluble in most organic solvents | |
100 mL | |
azane | |
6°C | |
CHEBI:16134 | |
AcroSeal™ Glass Bottle | |
Liquid |
Safety and Handling
- Ammonia, 2M solution in methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H315-Causes skin irritation.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only