missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, ca. 7N solution in methanol

Product Code. 10761394 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Plastic Can
Unit Size:
1 L
2.5 L
25 L
25mL
This item is not returnable. View return policy

Product Code. 10761394

Brand: Thermo Scientific Acros 133710025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ammonia, H3N, CAS Number-7664-41-7, 67-56-1, am-fol, ammonia, ammonia anhydrous, ammonia, anhydrous, ammonia gas, ammoniak, ammoniakgas, anhydrous ammonia, nitro-sil, spirit of hartshorn, 2.5L, CHEBI:16134, Colorless to Yellow, 6.0 to 7.5N, 0.7700g/mL, 05,15, 14 deg.C, 17.03, GHS Signal Word: Danger | CAS: 7664-41-7 | H3N | 17.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ammonia
Color Colorless to Yellow
Name Note ca. 7N Solution in Methanol
CAS 7664-41-7, 67-56-1
Molecular Formula H3N
Linear Formula NH3
Concentration 6.0 to 7.5N
MDL Number MFCD00011418
Fieser 05,15
Density 0.7700g/mL
Merck Index 15, 488
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Quantity 2.5 L
SMILES N
IUPAC Name azane
Molecular Weight (g/mol) 17.03
Flash Point 14°C
PubChem CID 222
ChEBI CHEBI:16134
Formula Weight 17.03
Packaging Glass Bottle
Specific Gravity 0.77
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonia, ca 7N solution in methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.