Learn More
Ammonia Dilute R2 According to European Pharmacopoeia (EP), Reagecon
High-purity, stable product for European Pharmacopeia applications.
Brand: Reagecon 1004703
Code : A0
Additional Details : CAS Number : 1336-21-6 Weight : 0.98500kg Transport : UN number : 1760 Chem class : 8 Pack group : III
Description
- European Pharmacopoeia Reagent, Solution
- NIST traceable
- High purity
- Accurate, stable and reproducible
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Developed and tested for European Pharmacopoeia (EP) applications.
- For preparation of other reagents (4.1.1), use in identification reactions of ions and functional groups (2.3.1) and used as a reagent in the performance of tests outlined in several EP monographs.
Specifications
EP Testing | |
1 L | |
Amber Glass Winchester Bottle GL45 |
Safety and Handling
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
- MIXTURE LIST-Contains : ammonia