missing translation for 'onlineSavingsMsg'
Learn More

Hach Pacific Pty Ltd Ammonia (Nitrogen) Test Strips, 0-6.0 mg/L

Suitable for both lab and field testing, Hach Test Strips are easy to use and easy to read.

Brand:  Hach Pacific Pty Ltd 2755325

Product Code. 15922797

  • £25.45 / Pack of 25
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Suitable for both lab and field testing, Hach Test Strips are easy to use and easy to read. Ammonia (Nitrogen) is a product of microbiological decay of plant and animal protein. Its presence in raw surface waters usually indicates domestic or agricultural pollution. Above certain levels, it is toxic to fish. Contains 25 tests. Easy to use, disposable, and inexpensive A great way to obtain quick, quantitative answers in the field or in the lab

Easy to use, disposable, and inexpensive
A great way to obtain quick, quantitative answers in the field or in the lab
Specifications
Footnote: *mg/L unless otherwise noted, ppb = μg/L, ppm = mg/L.; gpg = grains per gallon; 1 gpg = 17.1 mg/L or 17.1 ppm.
Model: Test Strips
Number of tests: 25
Parameter: Ammonia, Nitrogen, low range - As NH3-N, For freshwater
Platform: Test Strip
Range: 0 - 6 ppm
Ship Wt. (lbs): 0.25
Smallest Increment Steps: Steps: 0, 0.25, 0.5, 1.0, 3.0, 6.0 ppm
Units: 25 tests

Specifications

Specifications

25 Tests
0 to 6 ppm
Water Testing Kit Supplies and Reagents
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers