missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia solution 0.1mol/L, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Specifications
CAS | 7664-41-7 |
Packaging | Bottle |
Quantity | 1000 mL |
Molecular Formula | H3N |
MDL Number | MFCD00011418 |
InChI Key | QGZKDVFQNNGYKY-UHFFFAOYSA-N |
SMILES | N |
IUPAC Name | ammonia |
Molecular Weight (g/mol) | 17.03 |
PubChem CID | 222 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Product Identifier
- Ammonia 1M Volumetric Solution
- Danger
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'