missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution, 25%, Extra Pure, SLR, d=0.91, Fisher Chemical
Click to view available options
2.5L, Plastic Coated Glass bottle
Description
2.5L, Plastic Coated Glass bottle

Specifications
Specifications
pH | 12 |
Color | Colorless |
Boiling Point | 38°C |
CAS | 1336-21-6 |
Melting Point | -57°C |
Molecular Formula | H5NO |
MDL Number | MFCD00066650 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
SMILES | N.O |
IUPAC Name | amine hydrate |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 14
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2527049 | Certificate of Analysis | 18/02/2025 | A/3320/PB21,A/3320/PB17,A/3320/PB15 |
Safety and Handling



Product Identifier
- Ammonium hydroxide, 25-30% solution in water
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Ammonium hydroxide,Ammonia
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
57Hours
19Minutes
0Seconds
13New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonia Solution, 25%, Extra Pure, SLR, d=0.91, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction