missing translation for 'onlineSavingsMsg'
Learn More
Ammonia Solution, 25%, Extra Pure, SLR, d=0.91, Fisher Chemical Product Code.: 12686667

Ammonia Solution, 25%, Extra Pure, SLR, d=0.91, Fisher Chemical

Product Code. 12686667
5L, HDPE plastic bottle
Click to view available options
5L, HDPE plastic bottle
This item is not returnable. View return policy

Product Code. 12686667

Brand: Fisher Chemical A/3320/PB21

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

pH 12
Color Colorless
Boiling Point 38°C
CAS 1336-21-6
Melting Point -57°C
Molecular Formula H5NO
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 5 L
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
Packaging HDPE plastic bottle
Vapor Density 0.59
Vapor Pressure 500 hPa at 20°C
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide, 25-30% solution in water
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ammonium hydroxide,Ammonia
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonia Solution, 25%, Extra Pure, SLR, d=0.91, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.