missing translation for 'onlineSavingsMsg'
Learn More

Ammonia solution 33%, BAKER, J.T. Baker™

Product Code. 10473561
Click to view available options
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Laboratory Reagent

Specifications

Boiling Point 32°C to 37.7°C (1013 hPa)
CAS 1336-21-6
Melting Point -60°C to -57.5°C
Assay Percent Range 30 to 33%
Molecular Formula H5NO
MDL Number MFCD00066650
Density 0.88 - 0.91 g/cm³ (20°C)
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Quantity 1 L
SMILES N.O
IUPAC Name ammonium hydroxide
Molecular Weight (g/mol) 35.05
Residue after Ignition max. 0.003%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'