missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution, 35%, for Electronic Use, d=0.88, Fisher Chemical™
£66.45 - £84.60
Chemical Identifiers
CAS | 1336-21-6 |
---|---|
Molecular Weight (g/mol) | 35.05 |
InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
IUPAC Name | amine hydrate |
SMILES | N.O |
Description
Chemical Identifiers
1336-21-6 | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
N.O |
35.05 | |
amine hydrate |
Specifications
12 | |
1336-21-6 | |
MFCD00066650 | |
N.O | |
35.05 | |
Liquid |
Colorless | |
H5NO | |
VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
amine hydrate | |
35.05g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Ammonia solution 35%
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Ammonium hydroxide,Ammonia