missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution, 35%, Extra Pure, SLR, d=0.88, Fisher Chemical™
29.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
H5NO, CAS Number-1336-21-6, 2.5L, Colorless, 35.05g/mol, 11418, Plastic Coated Glass bottle, 12, Liquid
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 12 |
Color | Colorless |
Quantity | 1 L |
Formula Weight | 35.05g/mol |
Packaging | Plastic Coated Glass bottle |
Physical Form | Liquid |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- Ammonia solution 35%
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Ammonium hydroxide,Ammonia
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'