missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Solution, 7N in Methanol, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless |
Physical Form | Liquid |
Quantity | 100 mL |
Linear Formula | NH3 |
UN Number | UN3286 |
Flash Point | 14°C (57.2°F) |
IUPAC Name | ammonia |
Formula Weight | 17.03g/mol |
Concentration or Composition (by Analyte or Components) | 7N in methanol |
Packaging | Glass Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'