missing translation for 'onlineSavingsMsg'
Learn More
Ammonia Solution, Strong, Multi-Compendial, N.F., J.T. Baker™ Product Code.: 15548654
compliance-icons compliance-icons compliance-icons

Ammonia Solution, Strong, Multi-Compendial, N.F., J.T. Baker™

Product Code. 15548654
2.5 L, Glass Bottle
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 15548654

Brand: Avantor J.T.Baker 973605

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 7664-41-7
Quantity 2.5 L
Molecular Formula H3N
MDL Number MFCD00011418
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILES N
IUPAC Name ammonia
Molecular Weight (g/mol) 17.03
PubChem CID 222
ChEBI CHEBI:16134
Formula Weight 17.03
Grade Multi-Compendial/NF
Packaging Glass Bottle
Specific Gravity 1L = 0.90kg
Chemical Name or Material Ammonia Solution, Strong
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonia Solution, Strong, Multi-Compendial, N.F., J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.