missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ammonium Acetate, Ultra Pure Grade
Ammonium Acetate, Ultra Pure Grade is used in the purification and precipitation of DNA and for protein crystallization.
£29.80 - £89.90
Specifications
Assay | ≥97% |
---|---|
Chemical Name or Material | Ammonium Acetate |
CAS | 631-61-8 |
Content And Storage | Room Temperature |
Density | 1.17 at 68° F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17138548
|
MP Biomedicals™
0219384890 |
500 g |
£89.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17128548
|
MP Biomedicals™
0219384880 |
100 g |
£29.80
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Ammonium acetate, ultra pure, is for applications which require tight control of elemental content.
- Ammonium acetate is a widely used reagent in molecular biology and chromatography.
- Its applications include the purification and precipitation of DNA and protein crystallization.
- Ammonium acetate is commonly used in HPLC and MS analysis of various compounds, such as oligosaccharides, proteins, and peptides.
- Ammonium acetate is also used for the nonaqueous capillary electrophoresis-mass spectrometry (NACE-MS) of lipophilic peptides and therapeutic drugs.
Specifications
≥97% | |
631-61-8 | |
1.17 at 68° F | |
237.2° F | |
Ultrapure | |
Crystalline Powder |
Ammonium Acetate | |
Room Temperature | |
77.1 | |
C2H7NO2 | |
unii-rre756s6q2 |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.