missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate, Certified AR for Analysis, Fisher Chemical
Brand: Fisher Chemical A/3440/60
60.64 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
1kg, Plastic powder jar

Specifications
631-61-8 | |
White | |
Plastic powder jar | |
13066 | |
ammonium acetate, acetic acid, ammonium salt, azanium acetate, ammoniumacetate, acetic acid ammonium salt, ammonium ethanoate, unii-rre756s6q2, aconh4, ch3coonh4, ch3co2nh4 | |
CC(=O)[O-].[NH4+] | |
77.083 | |
CHEBI:62947 | |
Solid |
114°C | |
7 | |
C2H7NO2 | |
1 kg | |
USFZMSVCRYTOJT-UHFFFAOYSA-N | |
azanium;acetate | |
517165 | |
77.08g/mol |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
- Acetic acid, ammonium salt
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
3Hours
21Minutes
50Seconds
10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium Acetate, Certified AR for Analysis, Fisher Chemical > 1kg, Plastic powder jar
Spot an opportunity for improvement?Share a Content Correction