missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Acetate, Optima™ LC/MS Grade, Fisher Chemical™ Product Code.: 11317490

Ammonium Acetate, Optima™ LC/MS Grade, Fisher Chemical™

Product Code. 11317490
50g
Click to view available options
50g
This item is not returnable. View return policy

Product Code. 11317490

Brand: Fisher Chemical A11450

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ammonium Acetate, Optima LC/MS Grade, Fisher Chemical, CAS: 631-61-8, White, Melting Point: 114 deg.C, Molecular Formula: C2H7NO2, pH: 7, Quantity: 50g, Formula Weight: 77.08g/mol, MDL Number: 13066, Physical Form: Solid

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 114°C
CAS 631-61-8
Color White
pH 7
Physical Form Solid
Quantity 50 g
Molecular Formula C2H7NO2
MDL Number 13066
InChI Key USFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES [NH4+].CC([O-])=O
IUPAC Name ammonium acetate
Molecular Weight (g/mol) 77.08
Formula Weight 77.08g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Acetate, Optima™ LC/MS Grade, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.