missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium acetate, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥98%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
10 kg
20 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 100 g |
Linear Formula | CH3CO2NH4 |
Beilstein | 4186741 |
Formula Weight | 77.08g/mol |
Percent Purity | ≥98% |
Grade | ACS/puriss. p.a. |
Residue after Ignition | ≤0.01% (as SO4) |
Chemical Name or Material | Ammonium Acetate, Reag. Ph. Eur. |
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'