missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Acetate (White Crystals), Fisher BioReagents
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
Description
Ammonium Acetate can be used to precipitate DNA from enzymatic reactions.
Specifications
Specifications
Melting Point | 114°C |
Color | White |
pH | 7.0 |
Physical Form | Solid |
Quantity | 1 kg |
Assay Percent Range | ≥97 % |
Linear Formula | CH3COONH4 |
Merck Index | 15, 492 |
Formula Weight | 77.08 |
Percent Purity | ≥97% |
Show More |
Safety and Handling
Product Identifier
- Acetic acid, ammonium salt
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'