missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Aluminum Sulfate Dodecahydrate, Reagent Grade, ≥99% (Titration), Honeywell™
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 93.5°C |
Color | White |
pH | 2.6 (at 25°C) |
Quantity | 1 kg |
Linear Formula | AlNH4(SO4)2 · 12H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 453.33g/mol |
Percent Purity | ≥99% (titration) |
Odor | Odorless |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'