Learn More
Ammonium benzoate, 99%, Thermo Scientific Chemicals
Description
Ammonium benzoate is used predominantly in industries which acts as a corrosion inhibitors and anti-scaling agents . It is also used as paint additives and coating additives. Other consumer uses include cleaning and furnishing Care products, paints and coatings, personal care products, water treatment products. Ammonium benzoate is also used as chemical analysis reagents, in electronics industry, used for the production of electrolytic capacitors and also used in pharmaceutical industries. Used as adhesives, metal rust treatment and rubber additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 192°C to 198°C (decomposition) |
Color | White |
Physical Form | Crystalline |
Quantity | 100 g |
Flash Point | 110°C (230°F) |
Merck Index | 14,496 |
Solubility Information | It is soluble in water,methanol insoluble in diethyl ether. |
Formula Weight | 139.15 |
Percent Purity | 99% |
Odor | Weak |
Show More |
RUO – Research Use Only