missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium bicarbonate, Honeywell™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
CAS | 1066-33-7 |
Quantity | 50 g |
Molecular Formula | C3H8N2O9Zn2 |
Linear Formula | NH4HCO3 |
MDL Number | MFCD00012138 |
UN Number | UN3077 |
Beilstein | 4329606 |
Synonym | ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327 |
InChI Key | QOPSSYZLVIENBR-UHFFFAOYSA-J |
SMILES | [NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Safety and Handling

Product Identifier
- Ammonium bicarbonate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : ammonium hydrogencarbonate
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Days
4Hours
0Minutes
23Seconds
29
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ammonium bicarbonate, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction