missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium bicarbonate, Puriss., meets analytical spec. of Ph.Eur., BP, E 503, 99-101%, Honeywell Fluka™
20.74 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Packaging:
Fiberboard Box
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | NH4HCO3 |
Beilstein | 4329606 |
Formula Weight | 79.06g/mol |
Percent Purity | 99 to 101% |
Grade | puriss. |
Packaging | Poly bottle |
Density | Vapor Density: 2.7 (vs air) |
Chemical Name or Material | Ammonium Bicarbonate, Meets analytical specification of Ph.Eur.BP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'