missing translation for 'onlineSavingsMsg'
Learn More
Ammonium Bisulfate, Purum p.a., ≥98.0% (T), Honeywell™ Product Code.: 15630920

Ammonium Bisulfate, Purum p.a., ≥98.0% (T), Honeywell™

Product Code. 15630920 Shop All Honeywell Research Chemicals Products
1 kg, Poly bottle
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
This item is not returnable. View return policy

Product Code. 15630920

Brand: Honeywell Research Chemicals 098481KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purum p.a.,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 350°C
Melting Point 121°C to 145°C
CAS 7803-63-6
Color Colorless
pH 1 (100g/L at 20°C)
Physical Form Crystalline
Quantity 1 kg
Molecular Formula H5NO4S
Linear Formula (NH4)HSO4
MDL Number MFCD00051073
UN Number UN2506
Synonym ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4
Solubility Information Soluble in water, 0.5g in 10mL
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
SMILES [NH4+].OS([O-])(=O)=O
IUPAC Name ammonium hydrogen sulfate
Molecular Weight (g/mol) 115.10
PubChem CID 24655
Formula Weight 115.11g/mol
Percent Purity ≥98.0% (T)
Grade purum p.a.
Packaging Poly bottle
Chemical Name or Material Ammonium Bisulfate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ammonium Bisulfate, Purum p.a., ≥98.0% (T), Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.