Learn More
Ammonium bromide, ACS reagent, Thermo Scientific Chemicals
£46.13 - £188.94
Chemical Identifiers
CAS | 12124-97-9 |
---|---|
Molecular Formula | BrH4N |
Molecular Weight (g/mol) | 97.94 |
MDL Number | MFCD00011419 |
InChI Key | SWLVFNYSXGMGBS-UHFFFAOYSA-N |
IUPAC Name | ammonium bromide |
SMILES | [NH4+].[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12153751
|
Thermo Scientific Acros
446315000 |
500 g | Plastic bottle |
£46.13
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12143751
|
Thermo Scientific Acros
446310025 |
2.5 kg | Plastic Bottle |
£188.94
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12124-97-9 | |
97.94 | |
SWLVFNYSXGMGBS-UHFFFAOYSA-N | |
[NH4+].[Br-] |
BrH4N | |
MFCD00011419 | |
ammonium bromide |
Specifications
235.0°C | |
12124-97-9 | |
NH4Br | |
15,502 | |
SWLVFNYSXGMGBS-UHFFFAOYSA-N | |
ammonium bromide | |
97.94 | |
Ammonium bromide |
452.0°C | |
BrH4N | |
MFCD00011419 | |
Solubility in water: 970g/L (25°C). Other solubilities: 1456g/L (100°C), freely soluble in alcohol and acetone, slightly soluble in ether, practically insoluble in ethyl acetate | |
[NH4+].[Br-] | |
97.94 | |
ACS Reagent |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 235-183-8
- Ammonium bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only