missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Carbonate, Certified AR for Analysis, Powder, Fisher Chemical™
120.00 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Specifications
Melting Point | 58°C |
Color | White |
pH | 9.4 |
Physical Form | Solid |
Quantity | 1 kg |
Formula Weight | 96.09g/mol |
Packaging | Plastic powder jar |
Vapor Pressure | 69 hPa at 20°C |
Safety and Handling
Product Identifier
- Ammonium carbonate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'