missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Carbonate, Extra Pure, Powder, SLR, Fisher Chemical
20.36 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Use promo code "21743" to get your promotional price.
Click to view available options
500g, Plastic powder jar
Specifications
Melting Point | 58°C |
CAS | 506-87-6 |
Color | White |
pH | 9.4 |
Physical Form | Solid |
Quantity | 500 g |
Molecular Formula | CH8N2O3 |
MDL Number | 10890 |
InChI Key | PRKQVKDSMLBJBJ-UHFFFAOYSA-N |
SMILES | N.N.OC(O)=O |
Show More |
Safety and Handling
Product Identifier
- Ammonium carbonate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'