missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Carbonate, puriss., Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 kg |
Linear Formula | (NH4)2CO3 |
Beilstein | 4253487 |
Formula Weight | 96.09g/mol |
Grade | puriss. |
Residue after Ignition | ≤0.05% (as SO4) |
Density | Vapor Density: 2.7 (vs air) |
Chemical Name or Material | Ammonium Carbonate, Meets analytical specification of NF, Ph.Franc.,FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'